Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate, Xenopus and zebrafish based on 100% sequence homology.
This antibody contains enough material to conduct 10 mini-Western blots.
Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cleft palate 1; GABA(A) receptor subunit beta-3; GABR B3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; Gamma-aminobutyric acid receptor beta 3; Gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3
Gene Aliases: A230092K12Rik; AW049585; beta3; Cp1; Gabrb-3; Gabrb3
UniProt ID: (Mouse) P63080, (Rat) P63079
Entrez Gene ID: (Mouse) 14402, (Rat) 24922
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support