Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of GRK1/2 only when phosphorylated at Tyr13.
The sensation of sight is the result of a cascade of events starting with the interaction of photoactivated rhodopsin with a protein called transducin. Rhodopsin kinase is a G-protein-coupled Ser/Thr kinase, also known as GRK1, which is a key element in the regulation of this cascade. Following phosphorylation by GRK1, arrestin is recruited to phospho-rhodopsin quenching its phototransductive activity by preventing further interaction with transducin. By breaking the cycle of phototransduction, GRK1 plays an important role in the restoration of the system for subsequent visual events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; Beta-ARK-1; G protein-coupled receptor kinase 1; G protein-coupled receptpr kinase 1; G-protein coupled receptor kinase 2; G-protein receptor kinase 1; G-protein-coupled receptor kinase 2; grk 2; rhodopsin kinase; Rhodopsin kinase GRK1; RK
Gene Aliases: Adrbk-1; ADRBK1; BARK; Bark-1; BARK1; beta ARK; BETA-ARK1; betaARK1; GPRK1; GRK-2; GRK1; GRK2; RHOK; RK
UniProt ID: (Mouse) Q99MK8, (Rat) P26817, (Human) Q15835, (Human) P25098, (Rat) Q63651
Entrez Gene ID: (Mouse) 110355, (Rat) 25238, (Human) 6011, (Human) 156, (Mouse) 24013, (Rat) 81760
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support