Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of HAND1 only when phosphorylated at Ser98.
HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; heart and neural crest derivatives expressed transcript 1; Heart- and neural crest derivatives-expressed protein 1; Helix-loop-helix transcription factor expressed in extraembryonic mesoderm and trophoblast; Th1; Thing-1
Gene Aliases: BHLHA27; EHAND; Ehand1; HAND1; Hxt; Th1; Thing1
UniProt ID: (Human) O96004, (Mouse) Q64279, (Rat) P97832
Entrez Gene ID: (Human) 9421, (Mouse) 15110, (Rat) 59112
Molecular Function:
basic helix-loop-helix transcription factor
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support