Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of HBP1 only when phosphorylated at Ser380.
The HMG-box protein-1 (HBP1) is a member of the HMG family of transcription factors, which are characterized by the presence of a conserved protein motif, the high mobility group (HMG) 1 box, that mediates DNA binding. HBP1 binds to the tumor suppressor proteins Rb and p130 and initiates cell cycle arrest. Terminal cell differentiation requires this initial cell cycle arrest followed by the coordinated expression of genes defined as tissue-specific markers. Along with initiating the commitment to cell differentiation, the continued activity of HBP1 abrogates the expression of tissue-specific genes by associating with the MyoD proteins. In muscle cell differentiation, the MyoD family of transcription factors, which include Myf5, MyoD and myogenein, induce the expression of these cell-type specific proteins and contribute to the development of cell phenotypes. The progression of terminal differentiation is, therefore, dependent on both a decrease in HBP1 activity and the corresponding activation of MyoD-induced gene transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group box transcription factor 1; HMG box transcription factor 1; HMG box-containing protein 1
Gene Aliases: HBP1
UniProt ID: (Human) Q8TE93
Entrez Gene ID: (Human) 26959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support