Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of IGF1 Receptor only when phosphorylated at Tyr1146.
Insulin and Insulin-like Growth Factor receptors are transmembrane tyrosine kinase receptors that play critical roles in development, cell growth and metabolism. Insulin/Insulin-like growth factor-1 binding to the extracellular domain of IR and IGFR leads to autophosphorylation of the receptor and activation of the intrinsic tyrosine kinase activity, which allows appropriate substrates to be phosphorylated. Phosphorylation sites that are unique to each receptor presumably play a key role in these signaling differences. The catalytic loops within the tyrosine kinase domains contain a three tyrosine motif corresponding to Tyr1158, 1162 and 1163 (for the IR) and Tyr1131, 1135 and 1136 (for the IGF1R). Autophosphorylation of the tyrosine sites within the activation loop proceeds in a processive manner initiating at the second tyrosine (1162 or 1135), followed by phosphorylation at tyrosines 1158 (IGF-1R tyrosine 1131), 1163 (IGF-1R tyrosine 1136), resulting in the full activation of IR and IGF1R.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD220; CD221; IGF-1 receptor; IGF-I receptor; IGF-I Receptor beta; IGF1 Receptor; IGF1 Receptor beta; IGF1R beta; Insulin receptor; insulin receptor preproprotein; Insulin-like growth factor 1 receptor; insulin-like growth factor 1 receptor beta subunit; Insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; IR; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Aliases: 4932439J01Rik; A330103N21Rik; CD220; CD221; D630014A15Rik; D930020L01; HHF5; hyft; IGF-1R; IGF1R; IGFIR; IGFIRC; IGFR; Igfr1; INSR; IR; IR-A; IR-B; JTK13
UniProt ID: (Human) Q14CV2, (Human) P06213, (Rat) P24062, (Mouse) Q60751, (Mouse) P15208
Entrez Gene ID: (Pig) 397350, (Human) 3480, (Human) 3643, (Pig) 396755, (Rat) 25718, (Mouse) 16001, (Rat) 24954, (Mouse) 16337
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support