Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Antigen Affinity Purified from Pooled Serum
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral potassium-chloride cotransporter 2; Furosemide-sensitive K-Cl cotransporter; K-Cl cotransporter 2; KCC 2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; mKCC2; Neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; Solute carrier family 12 member 5; solute carrier family 12, member 5
Gene Aliases: KCC2; Kiaa1176; mKIAA1176; Slc12a5
UniProt ID: (Mouse) Q91V14, (Rat) Q63633
Entrez Gene ID: (Mouse) 57138, (Rat) 171373
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support