Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ZPK is a member of serine/threonine protein kinase family. This kinase contains a leucine-zipper domain, and is predominately expressed in neuronal cells. The phosphorylation state of this kinase in synaptic terminals was shown to be regulated by membrane depolarization via calcineurin. This kinase forms heterodimers with leucine zipper containing transcription factors, such as cAMP responsive element binding protein (CREB) and MYC, and thus may play a regulatory role in PKA or retinoic acid induced neuronal differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; Leucine-zipper protein kinase; M3KC; MAPK-upstream kinase; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); Mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; protein kinase MUK; Zipper (leucine) protein kinase; ZPK
Gene Aliases: DLK; MAP3K12; MEKK12; MUK; PK; ZPK; ZPKP1
UniProt ID: (Human) Q12852, (Rat) Q63796
Entrez Gene ID: (Human) 7786, (Rat) 25579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support