Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of MKP1/2 only when phosphorylated at Serine 296/318.
There are ten mitogen-activated protein kinase (MAPK) phosphatases (MKPs) that act as negative regulators of MAPK activity in mammalian cells. MKP2 (DUSP4) is a dual threonine/tyrosine phosphatase that specifically dephosphorylates and inactivates MAPKs (ERK, JNK, p38). It contains one Rhodanese domain and one tyrosine-protein phosphatase domain. MKP2 is expressed in a variety of tissues, and is localized in the nucleus. Transcription factor E2F-1 acts as a transcriptional regulator of MKP2. E2F-1/MKP2 pathway mediates apoptosis under oxidative stress and that MKP2 suppresses tumor formation. MKP2 is a novel transcription target of p53 in mediating apoptosis by 10-bp palindrome motif (CTGGCGCCAG) in the MKP2 promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3CH134/CL100 PTPase; CL 100; Dual specificity protein phosphatase 1; Dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH1; Dual specificity protein phosphatase hVH2; DUSP 4; DUSP-4; MAP kinase phosphatase 1; MAP kinase phosphatase 2; MAP kinase phosphatase-1; mitogen-activated protein (MAP) kinase phosphatase-1; Mitogen-activated protein kinase phosphatase 1; Mitogen-activated protein kinase phosphatase 2; mitogen-activated protein kinase phosphatase-1; MKP II; MKPII; oxidative stress-inducible protein tyrosine phosphatase; protein tyrosine phosphatase non-receptor type 16; protein tyrosine phosphatase, non-receptor type 16; Protein-tyrosine phosphatase 3CH134; Protein-tyrosine phosphatase CL100; Protein-tyrosine phosphatase ERP; Protein-tyrosine phosphatase non-receptor type 16; serine/threonine specific protein phosphatase; VH1 homologous phosphatase 2
Gene Aliases: 2700078F24Rik; 3CH134; AI844617; BB104621; CL100; DUSP1; DUSP4; E130306H24Rik; erp; HVH1; HVH2; MKP-1; MKP-2; MKP1; MKP2; PTPN10; Ptpn16; TYP; U19515; VH1; VH2
UniProt ID: (Human) P28562, (Human) Q13115, (Rat) Q64623, (Mouse) P28563, (Rat) Q62767, (Mouse) Q8BFV3
Entrez Gene ID: (Human) 1843, (Human) 1846, (Rat) 114856, (Mouse) 19252, (Rat) 60587, (Mouse) 319520
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support