Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-50; CENP-U; Centromere protein of 50 kDa; Centromere protein U; Interphase centromere complex protein 24; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; MLF1 interacting protein; MLF1-interacting protein; myeloid leukemia factor 1 interacting protein; Polo-box-interacting protein 1
Gene Aliases: 1700029A22Rik; CENP50; CENPU; CENPU50; ICEN24; KLIP1; MLF1IP; PBIP1
UniProt ID: (Human) Q71F23, (Rat) Q4V8G7, (Mouse) Q8C4M7
Entrez Gene ID: (Human) 79682, (Rat) 306464, (Mouse) 71876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support