Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HeLa. The target is usually found in the following locations: Cell membrane, Cytoplasm.
Immunogen sequence: TQNSIS
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mixed lineage kinase domain-like protein; mlkl
Gene Aliases: 9130019I15Rik; Mlkl
UniProt ID: (Mouse) Q9D2Y4
Entrez Gene ID: (Mouse) 74568
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support