Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Antibody detects endogenous levels of MOB1A only when phosphorylated at Tyr26.
MOB4A is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Mob4A regulates NDR1 and NDR2 kinase activity, widely expressed nuclear serine-threonine kinases that have been implicated in cell proliferation and/or tumor progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC33910; MOB kinase activator 1B; Mob1 homolog 1A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; Mob1A; Mob1B; Mps one binder kinase activator-like 1A
Gene Aliases: 1110003E08Rik; AU015450; B230364F10; MATS2; MOB1B; MOB4A; MOBKL1A
UniProt ID: (Human) Q7L9L4, (Mouse) Q8BPB0
Entrez Gene ID: (Human) 92597, (Rat) 360920, (Mouse) 68473
Molecular Function:
kinase activator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support