Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 10
Antibody detects endogenous levels of MITF only when phosphorylated at Serine 180.
Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, while the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHe32; black eyed white; Class E basic helix-loop-helix protein 32; microphtalmia-associated transcription factor; microphthalmia; Microphthalmia-associated transcription factor
Gene Aliases: BCC2; BHLHE32; Bw; CMM8; Gsfbcc2; MI; MITF; Vit; Vitiligo; Wh; WS2; WS2A
UniProt ID: (Human) O75030, (Rat) O88368, (Mouse) Q08874
Entrez Gene ID: (Human) 4286, (Rat) 25094, (Mouse) 17342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support