Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of NEDD4L only when phosphorylated at Ser342.
NEDD4L is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. NEDD4L inhibits TGF-beta signaling by triggering SMAD2 and TGFR1 ubiquitination and proteasome-dependent degradation. NEDD4L promotes ubiquitination and internalization of various plasma membrane channels such as ENaC, Nav1.2, Nav1.3, Nav1.5, Nav1.7, Nav1.8, Kv1.3, EAAT1 or CLC5. NEDD4L also promotes ubiquitination and degradation of SGK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase NEDD4-like; FLJ33870; HECT-type E3 ubiquitin transferase NED4L; Nedd4-2; NEDD4.2; neural precursor cell expressed, developmentally down-regulated gene 4b; ubiquitin-protein ligase Rsp5
Gene Aliases: 1300012C07Rik; hNEDD4-2; KIAA0439; NEDD4-2; NEDD4.2; Nedd4b; NEDD4L; NEDL3; RSP5
UniProt ID: (Human) Q96PU5
Entrez Gene ID: (Human) 23327, (Rat) 291553, (Mouse) 83814
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support