Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686J1293; FLJ21220; neurofibromatosis 1; Neurofibromatosis type 1; Neurofibromatosis-related protein NF-1; Neurofibromin
Gene Aliases: AW494271; E030030H24Rik; Nf-1; NF1; NFNS; VRNF; WSS
UniProt ID: (Human) P21359, (Mouse) Q04690, (Rat) P97526
Entrez Gene ID: (Human) 4763, (Mouse) 18015, (Rat) 24592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support