Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of NFAT5 only when phosphorylated at Ser145.
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutamine rich protein H65; NF-AT5; NFAT-like protein 1; Nuclear factor of activated T-cells 5; osmotic response element-binding protein; Rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; tonicity-responsive enhancer binding protein; Tonicity-responsive enhancer-binding protein
Gene Aliases: AI225870; B130038B15Rik; CAG-8; CAG80; KIAA0827; mKIAA0827; NF-AT5; NFAT5; NFATL1; NFATZ; OREBP; TONEBP
UniProt ID: (Human) O94916, (Mouse) Q9WV30
Entrez Gene ID: (Human) 10725, (Mouse) 54446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support