Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May sustain the structure of the postsynaptic density (PSD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; mNIFK; Nucleolar phosphoprotein Nopp34; Nucleolar protein interacting with the FHA domain of pKI-67
Gene Aliases: AI852665; C130020J04Rik; MKI67IP; NIFK; NOPP34
UniProt ID: (Human) Q9BYG3, (Mouse) Q91VE6
Entrez Gene ID: (Human) 84365, (Mouse) 67949
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support