Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
MA5-31847 has been tested in indirect ELISA.
NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARD, LRR, and NACHT-containing protein; caspase recruitment domain family, member 12; Caspase recruitment domain-containing protein 12; CED-4-like protein Clan; Ice protease-activating factor; ICE-protease activating factor; Ipaf; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4
Gene Aliases: 9530011P19Rik; AIFEC; CARD12; CLAN; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; IPAF; NLRC4; UNQ6189/PRO20215
UniProt ID: (Human) Q9NPP4, (Mouse) Q3UP24
Entrez Gene ID: (Human) 58484, (Mouse) 268973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support