Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
In mammals, there are several identified isoforms of p21-Activated Protein Kinases or PAKs: alpha-PAK (also known as PAK-1) and beta-PAK (also known as PAK-3) are mostly brain-specific, while gamma-PAK (also known as PAK-2) is expressed ubiquitously (Jakobi et al., 2003). Mutations of the gene coding for PAK-3 are associated wit X-linked mental retardation and recent work indicates that PAK-3 is a key regulator of synapse formation and plasticity in the hippocampus (Boda et al., 2004). PAK-3 is thought to play a key role in regulation of cell shape and motility as well as cell death (Jakobi et al., 2003; Walter et al., 1998). Autophosphorylation of Thr402 in the protein has been found to be essential for activation of PAK (Jakobi et al., 2000).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activated protein kinase alpha; ADRB2; adriamycin resistance-associated; Alpha-PAK; Beta-PAK; CDC42/RAC effector kinase PAK-A; CDC42/RAC effector kinase PAK-B; CDKN1A; EC 2.7.11.1; gamma pak; Gamma-PAK; hPAK3; kinase PAK1; Oligophrenin-3; p21 (CDKN1A)-activated kinase 1; p21 (CDKN1A)-activated kinase 2; p21 (CDKN1A)-activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 1; p21 protein (Cdc42/Rac)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 3; p21-activated kinase 1; p21-activated kinase 2; p21-activated kinase 3; p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast); p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p58; p65-PAK; p68-PAK; PAK 1; PAK alpha; PAK beta; PAK gamma; PAK-1; PAK-2; PAK-3; PAK65; Protein kinase MUK2; S6/H4 kinase; Serine/threonine-protein kinase PAK 1; Serine/threonine-protein kinase PAK 2; Serine/threonine-protein kinase PAK 3; STE20 homolog, yeast
Gene Aliases: 5330420P17Rik; A130002K10Rik; AI836325; ARA; AW045634; beta-PAK; bPAK; D16Ertd269e; gamma-PAK; mKIAA4182; MRX30; MRX47; OPHN3; PAK-1; PAK-2; PAK-3; PAK1; PAK2; PAK3; PAK3beta; PAK65; Pak65alpha; Pak65beta; Paka; PAKalpha; Pakb; PAKgamma; Stk4
UniProt ID: (Human) Q13153, (Human) Q13177, (Human) O75914, (Rat) P35465, (Mouse) Q8CIN4, (Rat) Q64303, (Mouse) O88645, (Rat) Q62829
Entrez Gene ID: (Human) 5058, (Human) 5062, (Human) 5063, (Mouse) 18479, (Rat) 29431, (Mouse) 224105, (Rat) 29432, (Mouse) 18481, (Rat) 29433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support