Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: C6, C6+PMA. The target is usually found in the following locations: Cytoplasm.
Immunogen sequence: IPSAK
PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15 kDa phosphoprotein enriched in astrocytes; Astrocytic phosphoprotein PEA-15; homolog of mouse MAT-1 oncogene; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; OTTHUMP00000029887; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; Phosphoprotein enriched in diabetes
Gene Aliases: HMAT1; HUMMAT1H; MAT1; MAT1H; PEA-15; PEA15; Pea15a; PED; Pkcs15
UniProt ID: (Human) Q15121, (Rat) Q5U318, (Mouse) Q62048
Entrez Gene ID: (Human) 8682, (Rat) 364052, (Mouse) 18611
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support