Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of PKA alpha CAT only when phosphorylated at Ser339.
Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C(s); C-alpha-2; C-alpha-S; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; MGC102831; MGC48865; PKA; PKA C-alpha; pkac; protein kinase A; protein kinase A catalytic subunit; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; sperm cAMP-dependent protein kinase catalytic subunit
Gene Aliases: Cs-PKA; PKACA; PKCA1; PKCD; PPNAD4; PRKACA
UniProt ID: (Human) P17612, (Mouse) P05132
Entrez Gene ID: (Human) 5566, (Rat) 25636, (Mouse) 18747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support