Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14 - 3 - 3 - zeta isoform; atypical protein kinase C; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; protein kinase C zeta subspecies; Protein kinase C zeta type; protein kinase C, zeta
Gene Aliases: 14-3-3-zetaisoform; AI098070; aPKCzeta; C80388; nPKC-zeta; PKC-ZETA; PKC2; Pkcz; PRKCZ; r14-3-3; R74924; zetaPKC
UniProt ID: (Human) Q05513, (Rat) P09217, (Mouse) Q02956
Entrez Gene ID: (Human) 5590, (Rat) 25522, (Mouse) 18762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support