Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive test controls include: Jurkat.
Immunogen sequence: RSTQG
Myosin phosphatase is a protein complex comprised of three subunits: a catalytic subunit (PP1c-delta, protein phosphatase 1, catalytic subunit delta), a large regulatory subunit (MYPT, myosin phosphatase target) and small regulatory subunit (sm-M20). Two isoforms of MYPT have been isolated--MYPT1 and MYPT2, the first of which is widely expressed, and the second of which may be specific to heart, skeletal muscle, and brain. Each of the MYPT isoforms functions to bind PP1c-delta and increase phosphatase activity. [NCBI]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leukocyte receptor cluster (LRC) encoded novel gene 3; leukocyte receptor cluster (LRC) member 3; myosin binding subunit; Myosin phosphatase target subunit 1; Myosin phosphatase target subunit 2; myosin phosphatase, target subunit 1; myosin phosphatase, target subunit 2; Myosin phosphatase-targeting subunit 1; Myosin phosphatase-targeting subunit 2; myosin-binding subunit 85; PPP1R12C; Protein phosphatase 1 myosin-binding subunit of 85 kDa; Protein phosphatase 1 myosin-binding subunit p85; Protein phosphatase 1 regulatory subunit 12A; Protein phosphatase 1 regulatory subunit 12B; Protein phosphatase 1 regulatory subunit 12C; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12B; protein phosphatase 1, regulatory (inhibitor) subunit 12C; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1, regulatory subunit 12B; protein phosphatase 1, regulatory subunit 12C; Protein phosphatase myosin-binding subunit
Gene Aliases: LENG3; M130; MBS; MBS85; MYPT1; MYPT2; p84; p85; PP1bp55; PPP1R12A; PPP1R12B; PPP1R12C
UniProt ID: (Human) O14974, (Human) O60237, (Human) Q9BZL4
Entrez Gene ID: (Human) 4659, (Human) 4660, (Human) 54776
Molecular Function: phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support