Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-SRC3 (Ser857) Polyclonal Antibody detects endogenous levels of SRC3 only when phosphorylated at Ser857.
AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACTR; AIB-1; Amplified in breast cancer 1 protein; amplified in breast cancer-1 protein homolog; bHLHe42; CBP-interacting protein; Class E basic helix-loop-helix protein 42; MGC141848; NCoA-3; Nuclear receptor coactivator 3; OTTHUMP00000031716; pCIP; RAC-3; Receptor-associated coactivator 3; RP5-1049G16.3; SRC-3; Steroid receptor coactivator protein 3; Thyroid hormone receptor activator molecule 1; TRAM-1
Gene Aliases: 2010305B15Rik; ACTR; AIB-1; AIB1; AW321064; BHLHE42; CAGH16; CTG26; KAT13B; NCOA3; p/Cip; pCIP; RAC3; SRC-3; SRC3; TNRC14; TNRC16; TRAM-1; TRAM1
UniProt ID: (Human) Q9Y6Q9
Entrez Gene ID: (Human) 8202, (Mouse) 17979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support