Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Phospho-STK38/STK38L (Thr444, Thr442) Polyclonal Antibody detects endogenous levels of STK38/STK38L only when phosphorylated at Thr444, Thr442.
This gene encodes a member of the AGC serine/threonine kinase family of proteins. The kinase activity of this protein is regulated by autophosphorylation and phosphorylation by other upstream kinases. This protein has been shown to function in the cell cycle and apoptosis. This protein has also been found to regulate the protein stability and transcriptional activity of the MYC oncogene. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ndr Ser/Thr kinase-like protein; NDR1 protein kinase; NDR2 protein kinase; nuclear Dbf2-related 2; Nuclear Dbf2-related kinase 1; Nuclear Dbf2-related kinase 2; serine/threonine kinase 38-like; Serine/threonine-protein kinase 38; Serine/threonine-protein kinase 38-like; stk38; STK38L
Gene Aliases: 4930473A22Rik; 5830476G13Rik; 9530097A09Rik; AA617404; B230328I19; KIAA0965; NDR; NDR1; NDR2; Ndr54; RGD1564793; STK38; STK38L
UniProt ID: (Human) Q15208, (Human) Q9Y2H1, (Mouse) Q91VJ4, (Mouse) Q7TSE6
Entrez Gene ID: (Human) 11329, (Human) 23012, (Mouse) 106504, (Rat) 361813, (Mouse) 232533, (Rat) 691337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support