Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with zebrafish, Xenopus, finch and canine based on 100% sequence homology.
This antibody is specific for the ~120 kDa TAO2 phosphorylated at Ser181 in Western blots of rat hippocampal lysate.
TAOK2 (TAO1) is a serine/threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway. In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hKFC-C; Kinase from chicken homolog C; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; Prostate-derived sterile 20-like kinase 1; PSK-1; serine-threonine kinase; serine/threonine protein kinase TAO2; Serine/threonine-protein kinase TAO2; tao; TAO kinase 2TAO kinase 2; Thousand and one amino acid protein 2; Thousand and one amino acid protein kinase 2
Gene Aliases: 1110033K02Rik; B230344N16; KIAA0881; MAP3K17; mKIAA0881; PSK; PSK1; PSK1-BETA; TAO1; TAO2; TAOK2; UNQ2971/PRO7431
UniProt ID: (Human) Q9UL54, (Mouse) Q6ZQ29, (Rat) Q9JLS3
Entrez Gene ID: (Human) 9344, (Mouse) 381921, (Rat) 64666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support