Search Thermo Fisher Scientific
FIGURE: 1 / 1
Keep as concentrated solution.
Predicted reactivity: Mouse (100%), Rat (100%), Sheep (100%).
Positive Control: 293T, 293T (200 ng/ml EGF treatment for 0.5 hr).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.