Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-RBL2 (Ser639) Polyclonal Antibody detects endogenous levels of RBL2 only when phosphorylated at Ser639.
RBL2 (also known as p130) or Retinoblastoma-like protein 2 is a key regulator of entry into cell division. It is directly involved in heterochromatin formation by maintaining overall chromatin structure and (in particular) that of constitutive heterochromatin by stabilizing histone methylation. RBL2 recruits and targets histone methyltransferases KMT5B and KMT5C leading to epigenetic transcriptional repression. It controls histone H4 'Lys20' trimethylation. It probably also acts as a transcription repressor by recruiting chromatin-modifying enzymes to promoters. RBL2 serves as a potent inhibitor of E2F-mediated trans-activation and associates preferentially with E2F5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130 kDa retinoblastoma-associated protein; FLJ26459; P130; PPAR-alpha-interacting complex protein 128; PRB2; PRIC128; rb2 p130; RBR-2; retinoblastoma-like 2; Retinoblastoma-like protein 2; Retinoblastoma-related protein 2
Gene Aliases: P130; PRB2; RB2; RBL2; RBR-2
UniProt ID: (Human) Q08999, (Mouse) Q64700, (Rat) O55081
Entrez Gene ID: (Human) 5934, (Mouse) 19651, (Rat) 81758
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support