Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Antigen Affinity Purified from Pooled Serum
RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates several residues of the S6 ribosomal protein. p70 S6 kinase is predominantly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70 S6 kinase acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis. The kinase activity of p70 S6 kinase leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding p70 S6 kinase and overexpression are seen in some breast cancer cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG10539-PA; CG10539-PB; Dp70s6k; dS6 kinase; EC 2.7.11.1; kinase p70S6K; KS6B1; p70 p70-S6K; p70 ribosomal S6 kinase; p70 S6 kinase; p70 S6 kinase 1; p70S6 kinase; p70S6K kinase; p70S6K-alpha; p80-S6K 1; p85 p85S6K; protein kinase-like 64F; ribosomal S6 protein kinase; RPS6-p70-protein kinase; S6 kinase; S6-kinase; S6K kinase; S6k-PA; S6k-PB; S6kinase
Gene Aliases: 10539; 7-10; 7084; CG10539; d-S6K; Dmel\CG10539; Dmel_CG10539; Dp70[s6k]; Dp70S6k; dps6k; DS6K; dS6K1; fs(3)07084; l(3)07084; p-S6K; p70 S6K; p70/S6K; p70[S6 kinase]; p70[S6K]; p70[S6kinase]; p70S6K; Pk64F; Pk?6; S6 K; S6K; S6K1; s6k11; S6k|S6K
Entrez Gene ID: (Fruit fly) 38654
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support