Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This monoclonal antibody detects two bands of 111 kDa and 55-60 kDa. The lower-molecular-weight band has been reported and likely represents a fragment resulting from cleavage of Pikachurin.
Immunogen sequence: MRFKTTAKD GLLLWRGDSP MRPNSDFISL GLRDGALVFS YNLGSGVASI MVNGSFNDGR WHRVKAVRDG QSGKITVDDY GARTGKSPGM MRQLNINGAL YVGGMKEIAL HTNRQYMRGL VGCISHFTLS TDYHISLVED SVDGKNINTC GAK (1-152 aa encoded by BC033177)
Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Agrin-like protein; EGF-like, fibronectin type III and laminin G domains; EGF-like, fibronectin type-III and laminin G-like domain-containing protein; Pikachurin
Gene Aliases: AGRINL; AGRNL; EGFLAM; PIKA
UniProt ID: (Human) Q63HQ2
Entrez Gene ID: (Human) 133584
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support