Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
Immunogen sequence: MAAGSRTSL LLAFALLCLP WLQEAGAVQT VPLSRLFDHA MLQAHRAHQL AIDTYQEFEE TYIPKDQKYS FLHDSQTSFC FSDSIPTPSN MEETQQKSNL ELLRISLLLI ESWLEPVRFL RSMFANNLVY DTSDSDDYHL LKDLEEGIQT LMGRLEDGSR RTGQILKQTY SKFDTNSHNH DALLKNYGLL YCFRKDMDKV ETFLRMVQCR SVEGSCGF (1-217 aa encoded by BC022044)
The human growth hormone (GH) locus is comprised by two GH (GH1 and GH2) genes and three chorionic somatomammotropin (CSH1, CSH2 and CSH-L) genes. Placental lactogens (CSH1, CSH2) are produced in the mammalian placenta and are similar in structure and function to growth hormones. Together, placental lactogens and growth factors play an essential role to assure successful lactation after pregnancy. Placental lactogens also modify the metabolic state of the mother during pregnancy to supply energy to the fetus. Both CSH1 and CSH2 can be detected by this antibody.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choriomammotropin; chorionic somatomammotropin A; chorionic somatomammotropin B; Chorionic somatomammotropin hormone 1; chorionic somatomammotropin hormone 1 (placental lactogen); Chorionic somatomammotropin hormone 2; chorionic somatomammotropin-1; growth hormone B1; growth hormone B3; Lactogen; PL; Placental lactogen
Gene Aliases: CS-1; CS-2; CSA; CSB; CSH1; CSH2; CSMT; GHB1; GHB3; hCS-1; hCS-A; hCS-B; PL
UniProt ID: (Human) P0DML3
Entrez Gene ID: (Human) 1442, (Human) 1443
Molecular Function: peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support