Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: plakophilin 2 in both locations: in the desmosomal plaque and in the karyoplasm. Plakophilin 2 is a member of the arm-repeat family of proteins which can occur in at least 2 splice forms (2a and 2b). Plakophilin 2 assembles with other proteins to form the desmosomal plaque in simple and glandular epithelia, the basal layer of certain stratified epithelia, all layers of some stratified epithelia, in Merkel cells, and desmosome-possessing nonepithelial cells such as myocardium, Purkinje fibers, and dendritic reticulum of lymph node follicles. In all these cells, including a variety of cell types devoid of desmosomes, plakophilin 2 is highly enriched in the nuclei (karyoplasm). Polypeptide reacting: Plakophilin 2, MW ~100 kD (SDS-PAGE); MW calculated from aa sequence: 92,750 (pI 9.33) and 97,410 (pI 9.38)Positive staining was obtained with several carcinomas derived from simple-type epithelia (e.g. hepatomas, colon carcinomas) and some adenocarcinomas derived from stratified epithelia (oesophagus carcinoma).Recommendation for immunohistochemistry on frozen sections: For better accessibility of the desmosome-associated antigen frozen sections might be pre-incubated for 5 min in PBS plus 0.2% Triton X-100 prior to antibody application. Longer incubation could result in a loss of Pkp2-nuclear staining. To prevent wash-out of soluble forms of the antigen, fixation with 2% formaldehyde is recommended.
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plakophilin-2
Gene Aliases: 1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l
UniProt ID: (Human) Q99959, (Mouse) Q9CQ73, (Rat) F1M7L9
Entrez Gene ID: (Human) 5318, (Mouse) 67451, (Rat) 287925
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support