Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VGSRADYDT LSLRSLRLGP GGLDDRYSLV SEQLEPAATS TYRAFAYERQ ASSSSSRAGG LDWPEATEVS PSRTIRAPAV RTLQRFQSSH RSRGVGGAVP GAVLEPVARA PSVRSLSLSL ADSGHLPDVH GFNSYGSHRT LQRLSSGFDD IDLPSAVKYL MASDPNLQVL GAAYIQHKCY SDAAAKKQAR SLQAVPRLVK LFNHANQEVQ RHATGAMRNL IYDNADNKLA LV (170-400 aa encoded by BC000081)
Plakophilin-3 is an Armadillo-like protein present in nuclei and desmosomes of epithelial cells. The expression pattern of this protein seems to be largely restricted to epithelial cell types. Plakophilin-3 can be detected along cell borders in a punctuate staining pattern typical for desmosomal proteins. In addition to the desmosomal immunolocalization, immunostaining was observed as bright nuclear speckles. Thus, like plakophilin-1 and-2, plakophilin-3 displays a dual intracellular localization in the desmosomal plaque and in the cell nucleus, and therefore is probably involved in signal transduction pathways between the plasma membrane and the nucleus. The human protein has a predicted molecular mass of ~87 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: plakophilin 3b; Plakophilin-3
Gene Aliases: 2310056L12Rik; PKP3
UniProt ID: (Human) Q9Y446, (Mouse) Q9QY23
Entrez Gene ID: (Human) 11187, (Mouse) 56460, (Rat) 293619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support