Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: GCLTQLYENA FFRGGDVASM YTPNAQYCQM RCTFHPRCLL FSFLPASSIN DMEKRFGCFL KDSVTGTLPK VHRTGAVSGH SLKQCGHQIS ACHRDIYKGV DMRGVNFNVS KVSSVEECQK RCTSNIRCQF FSYATQTFHK AEYRNNCLLK YSPGGTPTAI KVLSNVESGF SLKPCALSEI GCHMNIFQHL AFSDVDVARV LTPDAFVCRT ICTYHPNCLF FTFYTNVWKI ESQRNVCLLK TSESGTPSSS TPQENTISGY SLLTCKRTLP EPCHSKIYPG V; Positive Samples: Mouse liver, Mouse intestine, Mouse brain
Plasma prekallikrein is a glycoprotein that participates in the surface-dependent activation of blood coagulation, fibrinolysis, kinin generation and inflammation. It is synthesized in the liver and secreted into the blood as a single polypeptide chain. Plasma prekallikrein is converted to plasma kallikrein by factor XIIa by the cleavage of an internal Arg-Ile bond. Plasma kallikrein therefore is composed of a heavy chain and a light chain held together by a disulphide bond. The heavy chain originates from the amino-terminal end of the zymogen and contains 4 tandem repeats of 90 or 91 amino acids. Each repeat harbors a novel structure called the apple domain. The heavy chain is required for the surface-dependent pro-coagulant activity of plasma kallikrein. The light chain contains the active site or catalytic domain of the enzyme and is homologous to the trypsin family of serine proteases. Plasma prekallikrein deficiency causes a prolonged activated partial thromboplastin time in patients.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen, prostate specific; EC 3.4.21.34; Fletcher factor; kallikrein 3, plasma; kallikrein B plasma; kallikrein B, plasma (Fletcher factor) 1; Kininogenin; PKK; Plasma kallikrein; Plasma prekallikrein
Gene Aliases: APS; Kal-3; Kal3; Kalp1; KALP15; KLK3; KLKB1; Pk; PKK; PKKD; PPK; PSA
UniProt ID: (Human) P03952, (Mouse) P26262
Entrez Gene ID: (Human) 3818, (Rat) 25048, (Mouse) 16621
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support