Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody recognizes histidine rich protein II (HRP2) of P. falciparum.
Purity is >90%.
Recommended pairs for ELISA: MA5-18245 or MA5-18246 (capture) and MA5-18243 (conjugated, detection).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P. falciparum; P. falciparum HRP-2; P. falciparum HRP2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support