Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody raised against 1542-1841aa of human plexin B2 can recognize precursor and beta subunit of plexin B2.
Immunogen sequence: WKRVNTLMH YNVRDGATLI LSKVGVSQQP EDSQQDLPGE RHALLEEENR VWHLVRPTDE VDEGKSKRGS VKEKERTKAI TEIYLTRLLS VKGTLQQFVD NFFQSVLAPG HAVPPAVKYF FDFLDEQAEK HNIQDEDTIH IWKTNSLPLR FWVNILKNPH FIFDVHVHEV VDASLSVIAQ TFMDACTRTE HKLSRDSPSN KLLYAKEIST YKKMVEDYYK GIRQMVQVSD QDMNTHLAEI SRAHTDSLNT LVALHQLYQY TQKYYDEIIN ALEEDPAAQK MQLAFRLQQI AAALENKVTD L (1542-1841 aa encoded by BC004542)
Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved converting the single chain precursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2's possible role in the immune system has not been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: differentially expressed in brain tumors; MM1; MM1KIAA0315dJ402G11.3; Plexin-B2
Gene Aliases: Debt; dJ402G11.3; KIAA0315; MM1; Nbla00445; PLEXB2; plexin-B2; PLXNB2
UniProt ID: (Human) O15031, (Mouse) B2RXS4
Entrez Gene ID: (Human) 23654, (Mouse) 140570, (Rat) 315217
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support