Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPSPSPSPSQ NATQTTTDSS NKTAPTPASS VTIMATDTAQ QSTVPTSKAN EILASVKATT LGVSSDSPGT TTLAQQVSGP VNTTVARGGG SGNPTTTIES PKSTKSADTT TVATSTATAK PNTTSSQNGA EDTTNSGGKS SHSVTTDLTS TKAEHLTTPH PTSPLSPRQP TSTHPVATPT SSGHDHLMKI SSSSSTVAIP GYTFTSPGMT TTLPSSVISQ RTQQTSSQMP ASSTAPSSQE TVQPTSPATA LRTPTLPETM SSSPTAASTT HRYPKTPSPT VAHESNWAKC EDLETQTQSE KQLVLNLTGN TLCAGGASDE KLISLICRAV KATFNPAQDK CGIRLASVPG SQTVVVKEIT IHTKLPAKDV YERLKDKWDE LKEAGVSDMK LGDQGPPEEA EDRFSM
This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCTM-2 antigen; Gp200; MGC138240; PC; PCLP antibody; PCLP1 antibody; PODLX2; Podocalyxin; podocalyxin-like; Podocalyxin-like protein 1; podolyxin
Gene Aliases: Gp200; PC; PCLP; PCLP-1; PCLP1; PODXL
UniProt ID: (Human) O00592
Entrez Gene ID: (Human) 5420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support