Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
QED Bioscience
Reconstitute in 50 µL of distilled water (final concentration 2 mg/mL).
Collagen type I is present in soft connective tissue and bone where it represents approximately 90% of the organix matrix. Collagen type I is produced from procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. The propeptide levels can be measured in serum as indicators of collagen production. Elevated values of these propeptides in serum are found in patients with various bone metabolism defects. Procollagen type I C-terminal propeptide (PICP) has a molecular weight of approximately 100 kDa and the normal range in serum depends on age and sex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1 type I collagen; alpha1(I) procollagen; C-terminal Collagen; COL1A1; collagen alpha 1 chain type I; collagen alpha-1(I); Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen, type I, alpha 1; PICP; pro-alpha-1 collagen type 1; Procol; Procollagen; procollagen alpha 1 (I); type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen alpha 1 chain
Gene Aliases: COL1A1; EDSC; OI1; OI2; OI3; OI4
UniProt ID: (Dog) Q9XSJ7, (Human) P02452, (Bovine) P02453
Entrez Gene ID: (Dog) 403651, (Horse) 100033877, (Human) 1277, (Bovine) 282187
Molecular Function:
extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support