Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Expected to react with mouse samples due to 100% sequence homology.
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-neoendorphin; beta-neoendorphin; beta-neoendorphin-dynorphin; big dynorphin; dynorphin A(1-13); dynorphin A(1-17); dynorphin A(1-8); leu-enkephalin; leumorphin; preprodynorphin; proenkephalin B; proenkephalin-B; rimorphin
Gene Aliases: Dyn
Entrez Gene ID: (Mouse) 18610, (Rat) 29190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support