Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-14722 targets Prolactin in ELISA, IP, and RIA applications and shows reactivity with Human samples.
The MA5-14722 immunogen is human Prolactin.
MA5-14722 detects Prolactin which has a predicted molecular weight of approximately 25 kDa.
Product MA514722 is a smaller package size of MIP0202 (formerly sold as a Seradyn product).
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: decidual prolactin; growth hormone A1; Human prolactin; PRL; Prolactin
Gene Aliases: GHA1; PRL
UniProt ID: (Human) P01236
Entrez Gene ID: (Human) 5617
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support