Search Thermo Fisher Scientific
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-Pyranose oxidase antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Goat Serum as well as purified and partially purified Pyranose Oxidase [E.coli]. Cross reactivity against Pyranose Oxidase from other sources is unknown.
Pyranose Oxidase catalyzes the oxidation of various aldopyranoses and disaccharides on carbon-2 to the corresponding 2-keto sugars concomitant with the reduction of O2 to H2O2. It plays an important role in lignin degradation of wood rot fungi by supplying the essential cosubstrate H2O2 for the ligninolytic peroxidases, lignin peroxidase, and manganese-dependent peroxidase. The preferred substrate is D-glucose which is converted to 2-dehydro-D-glucose. Acts also on D-xylose, together with D-glucose the major sugars derived from wood, on L-sorbose, D-galactose and 1,5-anhydroglucitol, a diagnostic marker of diabetes mellitus.