Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RPQAGPLCLA GGASRAWGAP AGAAHPGPWG LWSSGR
Antigen sequence identity: Mouse 50%; Rat 44%
Pyrroline-5-carboxylate reductase (P5CR) is an enzyme that catalyzes the conversion of pyrroline-5-carboxylate (P5C) to proline, a non-essential amino acid that plays important roles in protein synthesis, cellular signaling, and stress response. P5CR is found in all living organisms and is essential for maintaining cellular homeostasis under various physiological and environmental conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support