Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: HepG2, HeLa, MCF7, A-549, U-251MG, Mouse liver, Mouse brain, Rat liver, Rat brain. The target is usually found in the following locations: Mitochondrion matrix.
Immunogen sequence: HSMGLGSKFK EVKKAYVEAN QMLGDLIKVT PSSKIVGDLA QFMVQNGLSR AEAEAQAEEL SFPRSVVEFL QGYIGVPHGG FPEPFRSKVL KDLPRVEGRP GASLPPLDLQ ALEKELVDRH GEEVTPEDVL SAAMYPDVFA HFKDFTATFG PLDSLNTRLF LQGPKIAEEF EVELERGKTL HIKALAVSDL NRAGQRQVFF ELNGQLRSIL VKDTQAMKEM HFHPKALKDV KGQIGAPMPG KVIDIKVVAG AKVAKGQPLC VLSAMKMETV VTSPMEGTVR KVHVTKDMTL EGDDLILEIE
Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.4.1.1; PCB; PYC; Pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; Pyruvic carboxylase
Gene Aliases: PC; PCB; Pcx
UniProt ID: (Human) P11498, (Rat) P52873
Entrez Gene ID: (Human) 5091, (Rat) 25104, (Mouse) 18563
Molecular Function:
ligase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support