Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-2 class I histocompatibility antigen, D-37 alpha chain; H2-Q1; MHC class I antigen Qa1; MHC classIb Qa1
Gene Aliases: 37b; 37c; H-2T23; H2-Qa1; H2-T23; Qa-1; Qa-1(b); Qa1; Qed-1; T18c; T18c(37); T23b; T23d
UniProt ID: (Mouse) P06339
Entrez Gene ID: (Mouse) 15040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support