Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAALDSLSLF TSLGLSEQKA RETLKNSALS AQLREAATQA QQTLGSTIDK ATGILLYGLA SRLRDTRRLS FLVSYIASKK IHTEPQLSAA LEYVRSHPLD PIDTVDFERE CGVGVIVTPE QIEEAVEAAI NRHRPQLLVE RYHFNMGLLM GEARAVLKWA DGKMIKNEVD MQVLHLLGPK LEADLEKKFK VAKARLEETD RRTAKDVVEN GETADQTLSL MEQLRGEALK FHKPGENYKT PGYVVTPHTM NLLKQHLEIT GGQVRTRFPP EPNGILHIGH AKAINFNFGY AKANNGICFL RFDDTNPEKE EAKFFTAICD MVAWLGYTPY KVTYASDYFD QLYAWAVELI RRGLAYVCHQ RGEELKGHNT LPSPWRDRPM EESLLLFEAM RKGKFSEGEA TLRMKLVMED GKMDPVAYRV KYTPHHRTGD KWCIYPTYDY THCLCDSIEH ITHSLCTKEF QARRSSYFWL CNALDVYCPV QWEYGRLNLH YAVVSKRKIL QLVATGAVRD WDDPRLFTLT ALRRRGFPPE AINNFCARVG VTVAQTTMEP HLLEACVRDV LNDTAPRAMA VLESLRVIIT NFPAAKSLDI QVPNFPADET KGFHQVPFAP IVFIERTDFK EEPEPGFKRL AWGQPVGLRH TGYVIELQHV VKGPSGCVES LEVTCRRADA GEKPKAFIHW VSQPLMCEVR LYERLFQHKN PEDPTEVPGG FLSDLNLASL HVVDAALVDC SVALAKPFDK FQFERLGYFS VDPDSHQGKL VFNRTVTLKE DPGKV
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GlnRS; Glutamine--tRNA ligase; glutamine-tRNA ligase; glutamine-tRNA synthetase; Glutaminyl-tRNA synthetase
Gene Aliases: 1110018N24Rik; 1200016L19Rik; C80286; GLNRS; MSCCA; PRO2195; QARS; QARS1; RGD1562301
UniProt ID: (Human) P47897, (Rat) Q66H61, (Mouse) Q8BML9
Entrez Gene ID: (Human) 5859, (Rat) 290868, (Mouse) 97541
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support