Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine
Glutamine-rich protein 1 contains 1 CARD domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20259; glutamine-rich 1; Glutamine-rich protein 1; MGC131838; Transcriptional regulator QRICH1
Gene Aliases: 2610028H07Rik; QRICH1
UniProt ID: (Human) Q2TAL8, (Mouse) Q3UA37
Entrez Gene ID: (Human) 54870, (Mouse) 69232
Molecular Function:
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support