Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RVVPKGKGE QIAREHGIRF FETSAKANIN IEKAFLTLAE DILRKTPVKE PNSENVDISS GGGVTGWKSK CC (130-200 aa encoded by BC000896)
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein RAB10; ras-related GTP-binding protein; Ras-related protein Rab-10
Gene Aliases: RAB10
UniProt ID: (Human) P61026
Entrez Gene ID: (Human) 10890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support