Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGTRDDEYDY LFKVVLIGDS GVGKSNLLSR FTRNEFNLES KSTIGVEFAT RSIQVDGKTI KAQIWDTAGQ ERYRAITSAY YRGAVGALLV YDIAKHLTYE NVERWLKELR DHADSNIVIM LVGNKSDLRH LRAVPTDEAR AFAEKNGLSF IETSALDSTN VEAAFQTILT EIYRIVSQKQ MSDRRENDMS PSNNVVPIHV PPTTENKPKV QCCQNI
Rab11A is a member of the YPT-1 subfamily of Ras-related GTP-binding proteins. Rab11A mRNA has been detected in a variety of tissues including brain, testis, spleen, heart, and gastrointestinal mucosa. In the gastric fundus the Rab11A protein is expressed in parietal cell tubulovesicles membranes. Expression of the Rab11A protein appears to occur in discrete epithelial cell populations where it localizes to apical vesicular populations. Recent studies have shown Rab11A is essential for resecretion of alpha-synuclein out of neuronal cells via endosome recycling and is required for exocytosis of discoidal/fusiform vesicles of bladder umbrella cells. Rab11A is also implicated in biogenesis of Birbeck granules and endosomal activation of the PI3K/AKT signaling pathway via G beta gamma trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAB 11A, member oncogene family; Rab-11; Ras-related protein Rab-11A; YL8
Gene Aliases: RAB11; RAB11A; YL8
UniProt ID: (Human) P62491
Entrez Gene ID: (Human) 8766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support