Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MGTRDDEYDY LFKVVLIGDS GVGKSNLLSR FTRNEFNLES KSTIGVEFAT RSIQVDGKTI KAQIWDTAGQ ERYRRITSAY YRGAVGALLV YDIAKHLTYE NVERWLKELR DHADSNIVIM LVGNKSDLRH LRAVPTDEAR AFAEKNNLSF IETSALDSTN VEEAFKNILT EIYRIVSQKQ IADRAAHDES PGNNVVDISV PPTTDGQKPN KLQCCQNL
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 11 is a known marker for protein trafficking, sorting, and recycling in the endosomal pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein YPT3; RAB11B, member of RAS oncogene family; Ras-related protein Rab-11B
Gene Aliases: H-YPT3; RAB11B; YPT3
UniProt ID: (Human) Q15907
Entrez Gene ID: (Human) 9230
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support