Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMLSEQAQK WFPTHVQVTV LQAKDLKPKG KSGTNDTYTI IQLGKEKYST SVAEKTLEPV WKEEASFELP GLLIQGSPEK YILFLIVMHR SLVGLDKFLG QVAINLNDIF EDKQRRKTEW FRLESKQGKR IKNRGEIKVN IQFMRNNMTA SMFDLSMKDK TRSPFAKLKD KMKGRKNDGT FSDTSSAIIP STHMPDANSE FSSGEIQMKS KPKKPFLLGP QRLSSAHSMS DLSGSHMSSE KLKAGTIGQT HLLGHQLDSF GTVPESGSLK SPHRRTLSFD TSKMNQPDSI VDEGELCFGR QNDPFTNVTA SLPQKFATLP RKKNPFEESS ETWDSSMNLF SKPIEIRKEN KREKREKVSL FERVTGKKDS RRSD (1-373 aa encoded by BC075073)
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NRip11; RAB11 family interacting protein 2 (class I); Rab11 family-interacting protein 2; rab11-family interacting protein 2; Rab11-FIP2; RAB11-FIP2 long isoform
Gene Aliases: 4930470G04Rik; A830046J09Rik; AW558126; KIAA0941; Nrip11; Rab11-FIP2; RAB11FIP2; RGD1308538
UniProt ID: (Human) Q7L804, (Mouse) G3XA57
Entrez Gene ID: (Human) 22841, (Rat) 308003, (Mouse) 74998
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support