Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
Rab effector involved in protein trafficking from apical recycling endosomes to the apical plasma membrane. Involved in insulin granule exocytosis. May regulate V-ATPase intracellular transport in response to extracellular acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gaf-1; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6
Entrez Gene ID: (Human) 26056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support